Key Documents
GN381080
Greiner CapMat for sealing multiwell plates
96 well, square nap, ethylene vinyl acetate, translucent
Synonym(s):
Greiner CapMats for sealing multiwell plates
About This Item
UNSPSC Code:
41122410
packaging
pack of 50 ea
manufacturer/tradename
Greiner 381080
availability
available only in China
Chem/Phys Resistance
Greiner CapMats are DMSO-resistant and withstand temperatures from -20°C to +60°C
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)