Skip to Content
Merck
All Photos(1)

Key Documents

G8653

Sigma-Aldrich

L-Glutamic acid γ-benzyl ester

Synonym(s):

L-Glutamic acid 5-benzyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OCOCH2CH2CH(NH2)COOH
CAS Number:
Molecular Weight:
237.25
Beilstein:
1885646
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97% (TLC)

form

powder

color

white to off-white

mp

181-182 °C (lit.)

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

N[C@@H](CCC(=O)OCc1ccccc1)C(O)=O

InChI

1S/C12H15NO4/c13-10(12(15)16)6-7-11(14)17-8-9-4-2-1-3-5-9/h1-5,10H,6-8,13H2,(H,15,16)/t10-/m0/s1

InChI key

BGGHCRNCRWQABU-JTQLQIEISA-N

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service