Skip to Content
Merck
All Photos(1)

Documents

G3751

Sigma-Aldrich

Glu-Val-Phe

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H27N3O6
CAS Number:
Molecular Weight:
393.43
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (TLC)

form

powder

color

white

storage temp.

−20°C

SMILES string

CC(C)C(NC(=O)C(N)CCC(O)=O)C(=O)NC(Cc1ccccc1)C(O)=O

InChI

1S/C19H27N3O6/c1-11(2)16(22-17(25)13(20)8-9-15(23)24)18(26)21-14(19(27)28)10-12-6-4-3-5-7-12/h3-7,11,13-14,16H,8-10,20H2,1-2H3,(H,21,26)(H,22,25)(H,23,24)(H,27,28)

InChI key

FVGOGEGGQLNZGH-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Bohley et al.
Biomedica biochimica acta, 50(4-6), 343-346 (1991-01-01)
Cellular proteins may be designated to fast degradation by their N-terminal amino acids, and especially a N-terminal arginine residue should have an extremely destabilizing effect on cytosol proteins. We investigated the post-translational arginylation of cytosol proteins and especially of ornithine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service