Skip to Content
Merck
All Photos(1)

Documents

EP5810718309

Rotor Key For Eppendorf® Rotors

for A-4-81 or A-4-81 -MTP/Flex on Models 5810 and 5810R

Synonym(s):

Rotor Key For Eppendorf® Rotors

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103909
NACRES:
NB.82

feature

for A-4-81 or A-4-81 -MTP/Flex on Models 5810 and 5810R

manufacturer/tradename

Eppendorf® 5810718309 (Europe)

technique(s)

centrifugation: suitable

Looking for similar products? Visit Product Comparison Guide

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service