Skip to Content
Merck
All Photos(1)

Documents

BPAEC

86123101, bovine lung, Endothelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

BPAEC, 86123101

biological source

bovine lung

growth mode

Adherent

karyotype

Not specified

morphology

Endothelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Bovine lung endothelium

Cell Line Description

BPAEC is susceptible to bovine respiratory viruses and is positive for BVDV.

Application

Virus studies: bovine respiratory viruses

Culture Medium

DMEM or 199 + 2mM Glutamine + 15% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:4 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service