Skip to Content
Merck
All Photos(1)

Key Documents

C1133

Sigma-Aldrich

Cytidine 3′-monophosphate

~98%

Synonym(s):

3′-Cytidylic acid, 3′-CMP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H14N3O8P
CAS Number:
Molecular Weight:
323.20
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:

Assay

~98%

storage temp.

−20°C

SMILES string

NC1=NC(=O)N(C=C1)[C@@H]2O[C@H](CO)[C@@H](OP(O)(O)=O)[C@H]2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juanjuan Ou et al.
Nature communications, 10(1), 1078-1078 (2019-03-08)
The efficacy of Fluorouracil (FU) in the treatment of colorectal cancer (CRC) is greatly limited by drug resistance. Autophagy has been implicated in chemoresistance, but the role of selective autophagic degradation in regulating chemoresistance remains unknown. In this study, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service