Skip to Content
Merck
All Photos(1)

Key Documents

B4758

Sigma-Aldrich

N-Benzoyl-Val-Gly-Arg p-nitroanilide hydrochloride

protease substrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H34N8O6 · HCl
CAS Number:
Molecular Weight:
591.06
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (TLC)

form

powder

solubility

ethanol: 20 mg/mL, clear, faintly yellow

storage temp.

−20°C

SMILES string

Cl.CC(C)C(NC(=O)c1ccccc1)C(=O)NCC(=O)NC(CCCNC(N)=N)C(=O)Nc2ccc(cc2)N(=O)=O

InChI

1S/C26H34N8O6.ClH/c1-16(2)22(33-23(36)17-7-4-3-5-8-17)25(38)30-15-21(35)32-20(9-6-14-29-26(27)28)24(37)31-18-10-12-19(13-11-18)34(39)40;/h3-5,7-8,10-13,16,20,22H,6,9,14-15H2,1-2H3,(H,30,38)(H,31,37)(H,32,35)(H,33,36)(H4,27,28,29);1H

InChI key

MLHRHUKHWCVSAU-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service