Skip to Content
Merck
All Photos(1)

Key Documents

65497

Sigma-Aldrich

TRIS acetate-EDTA Buffer solution

for Northern and Southern blotting, solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161700

grade

for molecular biology

product line

BioReagent

form

solution

impurities

DNases, none detected
RNases, none detected
phosphatases, none detected
proteases, none detected

pH

8.3±0.2

density

1.00 g/mL at 20 °C

storage temp.

room temp

General description

TBE (Tris/Acetate/EDTA) Buffer is commonly used in nucleic acid electrophoresis. This solution has a lower buffering capacity than TBE buffer, but dsDNA runs faster with TAE buffer. This concentrate can be easily diluted to 0.5x before use (with molecular biology grade water).

Application

Suitable for use as running buffer for nucleic acid electrophoresis, which may be part of Northern and Southern blot hybridization procedures. Especially useful for dsDNA separations.

Components

The TAE liquid buffer is a 1x solution that contains 0.04M TRIS acetate and 0.001M EDTA in water.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service