Skip to Content
Merck
All Photos(1)

Key Documents

23570

Sigma-Aldrich

Chlorobenzene

puriss. p.a., ACS reagent, ≥99.5% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5Cl
CAS Number:
Molecular Weight:
112.56
Beilstein:
605632
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.04
technique(s):
GC/GC: suitable
bp:
132 °C (lit.)
vapor pressure:
11.8 mmHg ( 25 °C)

grade

ACS reagent
puriss. p.a.

vapor density

3.86 (vs air)

vapor pressure

11.8 mmHg ( 25 °C)

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

1178 °F

expl. lim.

7.1 %

technique(s)

GC/GC: suitable

impurities

≤0.03% free acid (as CH3COOH)

evapn. residue

≤0.003%

refractive index

n20/D 1.524 (lit.)
n20/D 1.525

pH

7

bp

132 °C (lit.)

mp

−45 °C (lit.)

density

1.106 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.5 mg/kg
Ba: ≤0.1 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.1 mg/kg
K: ≤0.5 mg/kg
Li: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.1 mg/kg
Na: ≤0.5 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sr: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

SMILES string

Clc1ccccc1

InChI

1S/C6H5Cl/c7-6-4-2-1-3-5-6/h1-5H

InChI key

MVPPADPHJFYWMZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chlorobenzene is an aromatic compound that can be used as a solvent, degreasing agent, and chemical intermediate in various industries. It can be synthesized by the chlorination of benzene.

Application

Chlorobenzene can be used as a solvent in the:
  • Hydrosilylation of substituted benzaldehydes.
  • Knoevenagel condensation of aldehydes and ketones with alkyl nitriles.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

80.6 °F

Flash Point(C)

27 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Knoevenagel Condensation of Aldehydes and Ketones with Alkyl Nitriles Catalyzed by Strongly Basic Anion Exchange Resins under Continuous-Flow Conditions
Ishitani H, et al.
Asian Journal of Organic Chemistry, 7, 2061-2064 (2018)
Highly Efficient Large Bite Angle Diphosphine Substituted Molybdenum Catalyst for Hydrosilylation
Chakraborty S, et al.
ACS Catalysis, 3, 2208-2217 (2013)
Yiling Wang et al.
ACS applied materials & interfaces, 7(13), 7170-7179 (2015-03-21)
Solution-processed organic-inorganic hybrids composing of MoO3 nanoparticles and PEDOT:PSS were developed for use in inverted organic solar cells as hole transporting layer (HTL). The hybrid MoO3:PEDOT:PSS inks were prepared by simply mixing PEDOT:PSS aqueous and MoO3 ethanol suspension together. A
Sedlak DL and Andren AW.
Environmental Science & Technology, 25(4), 777-782 (1991)
Yang J, et al.
Macromolecules, 41(24), 9650-9655 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service