Skip to Content
Merck
All Photos(1)

Key Documents

90905

Supelco

3,3′-Di-O-methyl ellagic acid 4′-O-β-D-xylopyranoside

analytical standard

Synonym(s):

2-Hydroxy-3,8-dimethoxy-7-(β-D-xylopyranosyloxy)-[1]benzopyrano[5,4,3-cde][1]benzopyran-5,10-dione, 3,3′-Di-O-methyl-4-O-((β-D-xylopyranosyl)ellagic acid, 3-O-Methylducheside A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H18O12
CAS Number:
Molecular Weight:
462.36
Beilstein:
1612181
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥90.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

COc1c(O)cc2C(=O)Oc3c(OC)c(O[C@@H]4OC[C@@H](O)[C@H](O)[C@H]4O)cc5C(=O)Oc1c2-c35

InChI

1S/C21H18O12/c1-28-15-8(22)3-6-11-12-7(20(27)32-17(11)15)4-10(16(29-2)18(12)33-19(6)26)31-21-14(25)13(24)9(23)5-30-21/h3-4,9,13-14,21-25H,5H2,1-2H3/t9-,13+,14-,21+/m1/s1

InChI key

UDWUZPSSUIWBKB-LAPUEANGSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service