Skip to Content
Merck
All Photos(1)

Key Documents

70182

Millipore

Antibiotic Agar No 2

suitable for microbiology, NutriSelect® Plus

Synonym(s):

Penassay Base Agar, Grove and Randall Antibiotic Agar No. 2

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
RM 1,079.00

RM 1,079.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
500 G
RM 1,079.00

About This Item

UNSPSC Code:
41171504
NACRES:
NA.74

RM 1,079.00


Estimated to ship on29 May 2025


Request a Bulk Order

sterility

non-sterile

Quality Level

product line

BioChemika

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 15.0 g/L
meat extract, 1.5 g/L
peptone (from meat), 6.0 g/L
yeast extract, 3.0 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbe id | susceptibility testing: suitable
microbiological culture: suitable

final pH

6.5±0.2 (25 °C)

application(s)

clinical testing
food and beverages
life science and biopharma
pharmaceutical

microbiology

suitability

bacteria
nonselective for Bacillus spp.
nonselective for Staphylococcus spp.

Application

Used in antibiotic assay work (USP, FDA, Ph. Eur.) according to Grove & Randall; recommended as a base layer in the cylinder assay of e.g. penicillin and aureomycin, for the plate assay and the turbidimetric assay of bacitracin.

Preparation Note

Suspend 25.5 g in 1 litre of distilled water, heat to boiling to dissolve the medium completely. Sterilize in the autoclave at 121°C for 15 minutes.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D.C. Grove et al.
Assay Methods of Antibiotics, Med. Encycl. (1955)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service