Skip to Content
Merck
All Photos(1)

Key Documents

637696

Supelco

2-Chloro-1-(difluoromethoxy)-1,1,2-trifluoroethane-d2

98 atom % D, 98% (CP)

Synonym(s):

Enflurane-d2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCDFCF2OCF2D
Molecular Weight:
186.50
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

isotopic purity

98 atom % D

Assay

98% (CP)

bp

57 °C (lit.)

density

1.533 g/mL at 25 °C

mass shift

M+2

SMILES string

[2H]C(F)(F)OC(F)(F)C([2H])(F)Cl

InChI

1S/C3H2ClF5O/c4-1(5)3(8,9)10-2(6)7/h1-2H/i1D,2D

InChI key

JPGQOUSTVILISH-QDNHWIQGSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service