Skip to Content
Merck
All Photos(1)

Key Documents

60589

Supelco

Potassium tetraoxalate dihydrate

BioUltra, ≥99.5% (RT)

Synonym(s):

Oxalic acid hemipotassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KH3(C2O4)2 · 2H2O
CAS Number:
Molecular Weight:
254.19
Beilstein:
3854419
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

product line

BioUltra

Assay

≥99.5% (RT)

impurities

insoluble matter, passes filter test

pH

1.0-2.5 (25 °C, 0.1 M in H2O)

solubility

H2O: 0.1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤10 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

Al: ≤10 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
NH4+: ≤50 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

absorption

cut-off at 289 nm in H2O at 0.1 M

SMILES string

O.O.[K+].OC(=O)C(O)=O.OC(=O)C([O-])=O

InChI

1S/2C2H2O4.K.2H2O/c2*3-1(4)2(5)6;;;/h2*(H,3,4)(H,5,6);;2*1H2/q;;+1;;/p-1

InChI key

KPFHSTZUXXJAES-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Other Notes

Secondary standard buffer

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Data for Biochemical Research, 423-423 (1986)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service