Skip to Content
Merck
All Photos(1)

Documents

46834

Supelco

Sulfameter

VETRANAL®, analytical standard

Synonym(s):

5-Methoxysulfadiazine, N1-(5-Methoxypyrimidin-2-yl)sulfanilamide, Sulfamethoxydiazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12N4O3S
CAS Number:
Molecular Weight:
280.30
Beilstein:
621130
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

COc1cnc(NS(=O)(=O)c2ccc(N)cc2)nc1

InChI

1S/C11H12N4O3S/c1-18-9-6-13-11(14-7-9)15-19(16,17)10-4-2-8(12)3-5-10/h2-7H,12H2,1H3,(H,13,14,15)

InChI key

GPTONYMQFTZPKC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: sulfonamide
Sulfameter is a long-acting sulfonamide, which is found to be effective on urinary and respiratory tract infections. It is often used for animal disease treatment and prevention.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Sulfameter may be used as a reference standard in the determination of sulfameter in water samples and natural animal casings using solid phase extraction with high-performance liquid chromatography coupled with an ultraviolet detector (SPR-HPLC-UV).

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Studies of sulfamethoxydiazine.
Chew HW, et al.
Clinical Pharmacology and Therapeutics, 6(3), 307-315 (1965)
Analysis of sulfonamides in environmental water samples based on magnetic mixed hemimicelles solid-phase extraction coupled with HPLC-UV detection.
Sun L, et al.
Chemosphere, 77(10), 1306-1312 (2009)
Quantitative analysis of sulfonamide residues in natural animal casings by HPLC.
Sun H, et al.
Chromatographia, 66(5-6), 333-337 (2007)
Identification of polymorphs of sulfamethoxydiazine by optical and morphological methods.
T Yokoyama et al.
Chemical & pharmaceutical bulletin, 26(4), 1044-1048 (1978-04-01)
The effect of gastro-intestinal filling on the absorption of sulfametoxydiazine.
S Kaumeier et al.
International journal of clinical pharmacology and biopharmacy, 17(10), 412-415 (1979-10-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service