Skip to Content
Merck
All Photos(1)

Documents

05666

Supelco

Triclocarban

analytical standard

Synonym(s):

3,4,4′-Trichlorocarbanilide, 1-(4-Chlorophenyl)-3-(3,4-dichlorophenyl)urea, TCC, Triclocarban

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3NHCONHC6H4Cl
CAS Number:
Molecular Weight:
315.58
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor pressure

<0.1 mmHg ( 25 °C)

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

mp

254-256 °C (lit.)

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

Clc1ccc(NC(=O)Nc2ccc(Cl)c(Cl)c2)cc1

InChI

1S/C13H9Cl3N2O/c14-8-1-3-9(4-2-8)17-13(19)18-10-5-6-11(15)12(16)7-10/h1-7H,(H2,17,18,19)

InChI key

ICUTUKXCWQYESQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service