00718
1,1,1,2,2-Pentafluoro-4-iodobutane
≥99.0% (GC)
Synonym(s):
1H,1H,2H,2H-Perfluorobutyl iodide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
Assay
≥99.0% (GC)
refractive index
n20/D 1.391
bp
100-101 °C (lit.)
density
1.936 g/mL at 20 °C (lit.)
SMILES string
FC(F)(F)C(F)(F)CCI
InChI
1S/C4H4F5I/c5-3(6,1-2-10)4(7,8)9/h1-2H2
InChI key
PPXVPFPTMXFUDE-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Eye Irrit. 2
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service