Skip to Content
Merck

NM0046

NovaSeal Battery Charger

Battery Charger 100 -120 V US and Japan Plug

Synonym(s):

Battery charger for NovaSeal crimping tool

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41121815
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

sterility

non-sterile

product line

NovaSeal

compatibility

for use with NovaSeal battery powered crimping tool - NM0043



Still not finding the right product?

Explore all of our products under NovaSeal Battery Charger




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
NM004604054839045172