Skip to Content
Merck
All Photos(1)

Key Documents

MAVFLTFT1

Millipore

MAS-100 VF® Verification Report (Letter format)

The MAS-100 VF Sampler verification report is available to make verification faster and easier and ensures that your system′s performance remains compliant with its verificated specifications.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116500
eCl@ss:
25169090
NACRES:
JA.11

Quality Level

manufacturer/tradename

MAS-100®

input

sample type pharmaceutical(s)

application(s)

industrial qc

shipped in

ambient

Legal Information

MAS-100 is a registered trademark of MBV AG
MAS-100 VF is a registered trademark of MBV AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service