Skip to Content
Merck
All Photos(1)

Key Documents

MAB3836

Sigma-Aldrich

Anti-Green Fluorescent Protein Antibody

Chemicon®, from mouse

Synonym(s):

GFP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

monoclonal

species reactivity (predicted by homology)

all

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG

suitability

not suitable for immunohistochemistry

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Specificity

Reacts with Green Fluorescent Protein (GFP).

Immunogen

GFP

Application

Anti-Green Fluorescent Protein Antibody is an antibody against Green Fluorescent Protein for use in ELISA & WB.
Research Category
Epitope Tags & General Use
Research Sub Category
Epitope Tags
Western blot

ELISA

Not recommended for immunohistochemistry (for immunohistochemistry use MAB3580).

Optimal working dilutions must be determined by end user.

Physical form

Format: Purified
Purified immunoglobulin. Liquid in 0.02M Phosphate buffer containing 0.25M NaCl and 0.1% sodium azide.

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 6 months after date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Youjun Li et al.
Journal of Cancer, 7(12), 1645-1652 (2016-10-05)
Focal adhesions are large multi-protein complexes that serve as the linkage between extracellular matrix (ECM) and actin cytoskeleton and control the network of signaling cascades underlying cell migration. Talin plays a key role in focal adhesion turnover, and calpain-mediated proteolysis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service