Skip to Content
Merck
All Photos(2)

Documents

MAB1271-I

Sigma-Aldrich

Anti-Golgi Zone Antibody, clone 371-4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.74

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

371-4, monoclonal

mol wt

calculated mol wt 134.55 kDa

purified by

using protein G

species reactivity

human

species reactivity (predicted by homology)

monkey

packaging

antibody small pack of 100 μg

technique(s)

immunocytochemistry: suitable

isotype

IgG1κ

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

shipped in

2-8°C

target post-translational modification

unmodified

Gene Information

human ... GLG1(2734)

General description

Golgi apparatus protein 1 (UniProt: Q92896; also known as CFR-1, Cysteine-rich fibroblast growth factor receptor, E-selectin ligand 1, ESL-1, Golgi sialoglycoprotein MG-160) is encoded by the GLG1 (also known as CFR1, ESL1, MG160) gene (Gene ID: 2734) in human. The Golgi apparatus plays a major role in transport, processing, and sorting of thousands of macromolecules in eukaryotic cells. It is readily recognizable because of its stacked membrane-coated cavities known as dictyosomes. The Golgi complex consists of a stack of flattened, membrane-bound sacs known as cisternae, with three functional regions: the cis face, medial region and trans face and each of these regions contain various enzymes to selectively modify the macromolecules that pass through this complex. The Golgi complex is localized in the vicinity of the nucleus and is usually very large in secretory cells, where it fills most of the cytoplasm. Clone 371-4 selectively recognizes the Golgi complex in human cells. It produces a diffuse staining pattern of the Golgi zone in normal and malignant cells.

Specificity

Clone 371-4 is a mouse monoclonal antibody that detects Golgi apparatus.

Immunogen

SU-DHL-1 human large cell lymphoma cells.

Application

Quality Control Testing

Evaluated by Immunocytochemistry in HeLa cells.

Immunocytochemistry Analysis: A 1:500 dilution of this antibody detected Golgi apparatus protein 1 in HeLa cells.

Tested Application

Immunocytochemistry Analysis: A 1:500 dilution of this antibody detected Golgi apparatus protein 1 in A431cells.

Immunocytochemistry Analysis: A representative lot detected Golgi Zone in Immunocytochemistry applications (Endo, Y., et al. (2007). Cancer Sci.;98(10):1633-7; Strehlow, A.N., et al. (2007). Hum Mol Genet.;16(4):391-409; Oiso, S., et al. (2009). J Neurochem.;109(1):158-67; Leite, C., et al. (2014). PLoS One.;9(10):e111059).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-Golgi Zone, clone 371-4, Cat. No. MAB1271-I, is a mouse monoclonal antibody that detects Golgi apparatus protein 1 and is tested for use in Immunocytochemistry.

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommend storage at +2°C to +8°C. For long term storage antibodies can be kept at -20°C. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service