HCYTABA-17K
MILLIPLEX® Human Cytokine Autoantibody IgA Kit
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
species reactivity
human
Quality Level
manufacturer/tradename
Milliplex®
technique(s)
multiplexing: suitable
detection method
fluorometric (Luminex® xMAP®)
storage temp.
2-8°C
General description
Anti-cytokine antibodies occur frequently and are present in healthy individuals and patients with acquired immunodeficiency and autoimmune diseases. Cytokines offering protection against microbes can be targeted by cytokine autoantibodies, leading to life-threatening infections. Some infections associated with anti-cytokine antibodies include M. tuberculosis, Salmonella, S. aureus. E. coli, Histoplasma, and more. The presence of cytokine autoantibodies in autoimmune disease can influence disease severity and activity. These autoimmune diseases may include systemic lupus erythematosus, multiple sclerosis, Sjögren’s syndrome, rheumatoid arthritis, and acute respiratory distress syndrome. Measuring cytokine autoantibodies may be useful for disease monitoring and efficacy of treatment.
The MILLIPLEX® Human Cytokine Autoantibody IgA Magnetic Bead Panel is a 23-plex kit to be used for the simultaneous detection of any or all of the analytes listed below in serum and plasma samples:
The MILLIPLEX® Human Cytokine Autoantibody IgA Magnetic Bead Panel is a 23-plex kit to be used for the simultaneous detection of any or all of the analytes listed below in serum and plasma samples:
Application
The MILLIPLEX® Human Cytokine Autoantibody IgA Panel is a bead-based qualitative assay based on the Luminex® xMAP® technology, which enables the simultaneous analysis of cytokine autoantibodies in human serum and plasma samples.
Analytes Available:Anti-BAFF, Anti-G-CSF, Anti-IFNβ, Anti-IFNγ, Anti-IL-1α, Anti-IL-6, Anti IL 8, Anti-IL-10, Anti-IL-12p40, Anti-IL-15, Anti-IL-17A, Anti-IL-17F, Anti-IL-18, Anti-IL-22, Anti-TNFα, Anti-IFNα2, Anti-GM-CSF, Anti-IL-2, Anti-IL-3, Anti-IL-4, Anti-Osteopontin, Anti-PF4, and Anti-IFNω.
Panel type: MAGNETIC Immune Response
Cross Reactivity:There was no or negligible cross-reactivity between the antibodies for an analyte and any of the other analytes in this panel.
Research Category: Inflammation & Immunology
Research Sub-Category: Inflammation & Autoimmune Mechanisms; Inflammation
Analytes Available:Anti-BAFF, Anti-G-CSF, Anti-IFNβ, Anti-IFNγ, Anti-IL-1α, Anti-IL-6, Anti IL 8, Anti-IL-10, Anti-IL-12p40, Anti-IL-15, Anti-IL-17A, Anti-IL-17F, Anti-IL-18, Anti-IL-22, Anti-TNFα, Anti-IFNα2, Anti-GM-CSF, Anti-IL-2, Anti-IL-3, Anti-IL-4, Anti-Osteopontin, Anti-PF4, and Anti-IFNω.
Panel type: MAGNETIC Immune Response
Cross Reactivity:There was no or negligible cross-reactivity between the antibodies for an analyte and any of the other analytes in this panel.
Research Category: Inflammation & Immunology
Research Sub-Category: Inflammation & Autoimmune Mechanisms; Inflammation
Features and Benefits
Design your multiplex kit by choosing available analytes within this panel.
Packaging
96-well plate
Legal Information
Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Eye Irrit. 2 - Skin Sens. 1
Storage Class Code
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service