Skip to Content
Merck
All Photos(1)

Documents

CB1015

Sigma-Aldrich

Anti-GTAP Rabbit pAb

liquid, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

mouse, rat, human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

ambient

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... UBE2Q1(55585)

General description

Anti-GTAP, rabbit polyclonal, recognizes the ~67 kDa GTAP protein in HEK293 cells. It is validated for use in Western blotting.
Protein A purified rabbit polyclonal antibody. Recognizes the ~67 kDa GTAP protein.
Recognizes the ~67 kDa GTAP protein in HEK293 cells.
  • Hu Entrez ID: 55585
  • Mu Entrez ID: 70093
  • Rat Entrez ID: 295292
  • Target NCBI Accession: HP 060052.3
  • Target SwissProt: Q7Z7E8
  • The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes (E1s), ubiquitin-conjugating enzymes (E2s), and ubiquitin-protein ligases (E3s). GTAP is a member of the E2 ubiquitin-conjugating enzyme family. GTAP may participate in intercellular adhesion and embryonic development. GTAP is thought to play a role in stem cell development via its regulation of laminin-dependent embryonic cell adhesion and the formation of embryoid bodies.

    Immunogen

    A synthetic peptide corresponding to amino acids 7-22 (QGQQQPGPGQQLGGQG) of human GTAP, conjugated to KLH.
    Human

    Application


    Immunoblotting (3-10 g/ml)

    Warning

    Toxicity: Regulatory Review (Z)

    Physical form

    In PBS.

    Reconstitution

    Following initial thaw, aliquot and freeze (-20°C).

    Other Notes

    Due to a high degree of sequence identity, this antibody is also expected to react with mouse and rat GTAP. Antibody should be titrated for optimal results in individual systems.

    Legal Information

    CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

    Not finding the right product?  

    Try our Product Selector Tool.

    Storage Class Code

    12 - Non Combustible Liquids

    WGK

    WGK 2

    Flash Point(F)

    Not applicable

    Flash Point(C)

    Not applicable


    Certificates of Analysis (COA)

    Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service