8910-OP
TE Buffer Solution
OmniPur® Grade, 100X Liquid Concentrate, pH 8.0
Synonym(s):
TRIS-EDTA Buffer
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
form
liquid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
color
haze-free colorless
shipped in
ambient
storage temp.
15-25°C
General description
General Composition: Each liter of concentrate makes 100 L of 1X TE buffer (1X = 0.01 M TRIS, 0.001 M EDTA Na2). Molecular Biology grade.
Conductivity (1X concentration) at 25°C: 775-975 µmho
DNase: None detected
pH (1X concentration) at 25°C: 7.9-8.1
Protease: None detected
RNase: None detected
Intended for laboratory and manufacturing use only. Not for drug, food, or household use.
Application
- Evaluation of Microbial Profile in Patients with Polycystic Ovary Syndrome and Periodontal Disease: A Case-Control Study.: This study investigates the microbial profiles in patients with PCOS and periodontal disease. TE buffer solutions are utilized for the extraction and preservation of microbial DNA, ensuring high-quality samples for subsequent analysis and comparison (Achu Joseph et al., 2023).
- A dual paper-based nucleic acid extraction method from blood in under ten minutes for point-of-care diagnostics.: This paper introduces a rapid and efficient method for nucleic acid extraction from blood using a dual paper-based approach. TE buffer solutions play a crucial role in stabilizing and preserving nucleic acids during the extraction process, making the method suitable for point-of-care diagnostics (Malpartida-Cardenas et al., 2023).
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service