8.45042
Magnesium
turnings acc. to Grignard Msynth™plus
Synonym(s):
Magnesium
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
Mg
CAS Number:
Molecular Weight:
24.31
MDL number:
UNSPSC Code:
12141502
EC Index Number:
231-104-6
Recommended Products
form
chips
autoignition temp.
>450 °C
bp
1107 °C/1013 hPa
mp
651 °C
density
1.75 g/cm3 at 20 °C
storage temp.
2-30°C
InChI
1S/Mg/q+2
InChI key
JLVVSXFLKOJNIY-UHFFFAOYSA-N
Analysis Note
Color (visual): grey
Appearance of substance (visual): chips
Assay (on Mg): ≥ 99.0 %
Splinter: ≤ 4 mm
Pb (Lead): lot specific result
1,1-Dichloroethene (HS-GC): lot specific result
1,1,1-Trichloroethane (HS-GC): lot specific result
Carbon tetrachloride (HS-GC): lot specific result
Benzene (HS-GC): lot specific result
1,2-Dichloroethane (HS-GC): lot specific result
Appearance of substance (visual): chips
Assay (on Mg): ≥ 99.0 %
Splinter: ≤ 4 mm
Pb (Lead): lot specific result
1,1-Dichloroethene (HS-GC): lot specific result
1,1,1-Trichloroethane (HS-GC): lot specific result
Carbon tetrachloride (HS-GC): lot specific result
Benzene (HS-GC): lot specific result
1,2-Dichloroethane (HS-GC): lot specific result
Legal Information
MSYNTH is a trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Sol. 1 - Self-heat. 1 - Water-react. 2
Storage Class Code
4.2 - Pyrophoric and self-heating hazardous materials
WGK
nwg
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service