Skip to Content
Merck
All Photos(1)

Key Documents

8.22197

Sigma-Aldrich

Lithium tetrahydroborate

for synthesis

Synonym(s):

Lithium tetrahydroborate, Lithium boron hydride, Lithium boranate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
H4BLi
CAS Number:
Molecular Weight:
21.78
MDL number:
UNSPSC Code:
12352306
EC Index Number:
241-021-7

Quality Level

expl. lim.

4.00-75.60 % (v/v)

reaction suitability

reagent type: reductant

mp

280 °C (decomposition)

solubility

209 g/L

density

0.666 g/cm3 at 20 °C

bulk density

300 kg/m3

Analysis Note

Assay (hypochlorite): ≥ 93.0 %
Identity (ICP-OES): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Skin Corr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials, which set free flammable gases upon contact with water

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service