Skip to Content
Merck
All Photos(1)

Key Documents

8.18598

Sigma-Aldrich

2,3-Epoxypropyl isopropyl ether

for synthesis

Synonym(s):

2,3-Epoxypropyl isopropyl ether, Isopropoxymethyloxirane, Isopropyl glycidyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O2
CAS Number:
Molecular Weight:
116.16
MDL number:
UNSPSC Code:
12352112
EC Index Number:
223-672-9
Pricing and availability is not currently available.

vapor pressure

13 hPa ( 25 °C)

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

290 °C

potency

4200 mg/kg LD50, oral (Rat)
9650 mg/kg LD50, skin (Rabbit)

pH

7 (20 °C, 188 g/L in H2O)

bp

137 °C/1013 hPa

transition temp

flash point 33 °C

density

0.92 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C6H12O2/c1-5(2)7-3-6-4-8-6/h5-6H,3-4H2,1-2H3

InChI key

NWLUZGJDEZBBRH-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.918 - 0.920
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Flam. Liq. 3 - Muta. 2 - Resp. Sens. 1 - Skin Sens. 1

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

91.4 °F - closed cup

Flash Point(C)

33 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service