Skip to Content
Merck

8.17081

SAFC

Sodium caprylate

EMPROVE® EXPERT, Ph. Eur., ChP, NF

Pharma Manufacturing

Synonym(s):

Sodium octanoate, Sodium Caprylate, Octanoic Acid Sodium Salt, Caprylic acid sodium salt, Octanoic acid sodium salt, Sodium caprylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6COONa
CAS Number:
Molecular Weight:
166.19
Beilstein:
3597723
MDL number:
UNSPSC Code:
12352107
EC Index Number:
217-850-5

Agency

ChP
NF
Ph. Eur.

Quality Level

description

suitable for use as excipient EMPROVE® exp Ph Eur,ChP,NF

product line

EMPROVE® EXPERT

form

solid

pH

8.0-10.5 (20 °C, 100 g/L in H2O)

mp

>225 °C

application(s)

liquid formulation
parenterals
pharma/biopharma processes
pharmaceutical

storage temp.

2-30°C

SMILES string

[Na+].CCCCCCCC([O-])=O

InChI

1S/C8H16O2.Na/c1-2-3-4-5-6-7-8(9)10;/h2-7H2,1H3,(H,9,10);/q;+1/p-1

InChI key

BYKRNSHANADUFY-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Chemicals play an important role in the stabilization of a biologic drug during its manufacturing and formulation process – for instance, by preventing aggregation. We offer a wide range of high-quality stabilizers, buffers and salts to successfully purify and formulate your biomolecules. Specifically developed for high-risk applications, they are low in bioburden and endotoxins.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Sodium caprylate is widely used in purification of plasma derived therapeutics.
Due to its low microbial and endotoxin limits, Sodium caprylate Emprove® Expert is especially suitable for high-risk applications.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

The substance 8.17081 has no REACH registration because the only supported use is the use in medicinal products for human or veterinary use or in food or feedingstuffs according to article 2 of the REACH Regulation (EC) No 1907/2006.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service