Skip to Content
Merck
All Photos(1)

Key Documents

70034-M

Sigma-Aldrich

BacPlaque Agarose - Novagen

Qualified agarose for optimal plaque assays

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.54

Quality Level

form

solid

manufacturer/tradename

Novagen®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

storage temp.

10-30°C

General description

Agarose overlays are commonly used in baculovirus plaque assays. Many commercial DNA grade agaroses either contain contaminants that are toxic to insect cells or lack the physical characteristics required to obtain good overlays. Novagen BacPlaque<TMSYMBOL></TMSYMBOL> Agarose is pre-qualified in plaque assays and transfections to ensure consistent performance.
Qualified agarose for optimal plaque assays

Warning

Toxicity: Standard Handling (A)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service