Skip to Content
Merck
All Photos(1)

Key Documents

454861

Sigma-Aldrich

MNK1 Inhibitor

The MNK1 Inhibitor, also referenced under CAS 522629-08-9, controls the biological activity of MNK1. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

MNK1 Inhibitor, 4-Amino-5-(4-fluoroanilino)-pyrazolo[3,4-d]pyrimidine, CGP 57380, MNK Inhibitor I, Mitogen-Activated Protein Kinase-Interacting Kinase Inhibitor I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H9FN6
CAS Number:
Molecular Weight:
244.23
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 15 mg/mL
ethanol: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C11H9FN6/c12-6-1-3-7(4-2-6)16-11-8-9(13)14-5-15-10(8)17-18-11/h1-5H,(H4,13,14,15,16,17,18)

InChI key

UQPMANVRZYYQMD-UHFFFAOYSA-N

General description

A cell-permeable, reversible, and ATP-competitive pyrazolo-pyrimidine compound that acts as a selective inhibitor of mitogen-activated protein kinase-interacting kinase 1 (MNK1; IC50 = 2.2 µM) with no inhibitory activity against p38, JNK1, ERK1/2, PKC, or Src-like kinases. Effectively inhibits eIF4E phosphorylation (IC50 = 3 µM) by MNK1 in 293 human embryonic kidney cells and serves as a useful tool in studying MNK1-mediated cellular signaling.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Mitogen-activated protein kinase-interacting kinase 1 (MNK1)
Product competes with ATP.
Reversible: yes
Target IC50: 2.2 µM against mitogen-activated protein kinase-interacting kinase 1 (MNK1); 3 µM against eIF4E phosphorylation by MNK1 in 293 human embryonic kidney cells

Packaging

Packaged under inert gas

Warning

Toxicity: Toxic (F)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Topisirovic, I., et al. 2004. Cancer Res.64, 8639.
Worch, J., et al. 2004. Oncogene23, 9162.
Walsh, D., and Mohr, I. 2004. Genes Dev.18, 660.
Morley, S.J., and Naegele, S. 2002. J. Biol. Chem.277, 32855.
Knauf, U., et al. 2001. Mol. Cell. Biol.21, 5500.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benedetta De Ponte Conti et al.
eLife, 10 (2021-11-18)
We performed a systematic analysis of the translation rate of tumor-infiltrating lymphocytes (TILs) and the microenvironment inputs affecting it, both in humans and in mice. Measurement of puromycin incorporation, a proxy of protein synthesis, revealed an increase of translating CD4+

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service