Skip to Content
Merck
All Photos(1)

Key Documents

376820

Sigma-Aldrich

β-Hexosaminidase Inhibitor - CAS 281224-40-6 - Calbiochem

Synonym(s):

M-31850

Sign Into View Organizational & Contract Pricing


About This Item

Assay

≥95% (HPLC)

form

solid

color

white

solubility

DMSO: 1 mg/mL

General description

A cell-permeable bisnaphthalimide compound that acts as a substrate-competitive and selective inhibitor against human β-N-acetyl hexosaminidase (IC50 = 6.0 and 3.1 μM for hHex A and hHex B, respectively), while exhibiting much reduced potency against Jack Bean or bacterial Hex (IC50 = 280 & >500 μM for JBHex & SpHex, respectively) and little or no activity against SmHex or hOGN. Shown to function as a pharmacological chaperone and effectively enhance the transport of mutant Hex alpha subunit from the ER to lysosome in cells derived from ISD and ATSD patients.
The β-Hexosaminidase Inhibitor controls the biological activity of β-Hexosaminidase. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at 20°C.

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael B Tropak et al.
Chemistry & biology, 14(2), 153-164 (2007-02-24)
The adult forms of Tay-Sachs and Sandhoff diseases result when the activity of beta-hexosaminidase A (Hex) falls below approximately 10% of normal due to decreased transport of the destabilized mutant enzyme to the lysosome. Carbohydrate-based competitive inhibitors of Hex act

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service