Skip to Content
Merck
All Photos(1)

Documents

189733

Sigma-Aldrich

Streptavidin, Peroxidase Conjugate

This Streptavidin, Peroxidase Conjugate is validated for use in Enzyme Immunoassay.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116133
NACRES:
NA.54

Quality Level

form

liquid

contains

0.02% Thimerosal as preservative

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

shipped in

wet ice

storage temp.

2-8°C

General description

Streptavidin conjugated to horseradish peroxidase. Specific for biotin, biotinylated proteins, or biotinylated antibodies.

Application

Enzyme Immunoassay (1:10,000)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 10 mM PBS, 0.5 mM EDTA, 10% mannose, 2% sucrose, 1% BSA, pH 7.6.

Other Notes

Roth, J., et al. 1992. Histochemistry98, 229.
Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

To comply with ban of sale of mercury-added products required by The Interstate Mercury Education and Reduction Clearinghouse (IMERC), this product is prohibited to be sold in the following US states: Rhode Island and Connecticut.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juliane Schwaderer et al.
Cell death & disease, 11(2), 154-154 (2020-03-01)
Liver receptor homolog-1 (LRH-1, Nr5a2) is an orphan nuclear receptor mainly expressed in tissues of endodermal origin, where its physiological role has been extensively studied. LRH-1 has been implicated in liver cell differentiation and proliferation, as well as glucose, lipid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service