1.46780
Buffered NaCl-Peptone Solution
ready-to-use, bottle volume 200 mL , filling volume
Synonym(s):
Buffered NaCl Peptone Solution, Buffered Saline Peptone Water, Buffered Sodium Chloride Peptone Water, NaCl Peptone Solution (Buffered), Sodium Chloride Peptone Buffer
About This Item
Recommended Products
Agency
EP (2.6.13)
JP (4.06)
USP
Quality Level
description
For the microbiological examination of non-sterile pharmaceuticals
form
liquid
shelf life
12 mo.
feature
closure type screw cap with septum
ready-to-use
packaging
pkg of 200 mL (in 250 mL bottle with screw cap with septum)
pkg of 6 bottles (per box)
bottle of
technique(s)
direct inoculation: suitable
microbiological culture: suitable
sterile filtration: suitable
bottle capacity
250 mL
bottle volume
200 mL , filling volume
color
clear colorless
pH
6.8-7.2
application(s)
food and beverages
pharmaceutical
storage temp.
2-25°C
suitability
nonselective for
General description
Application
Features and Benefits
Analysis Note
Storage Class Code
10 - Combustible liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service