Skip to Content
Merck
All Photos(1)

Key Documents

144151

Carrying Case

for use with RCS® High Flow

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122409

compatibility

for use with RCS® High Flow

shipped in

ambient

storage temp.

room temp

General description

194326 (RCS High Flow)
Spare part, aluminum carrying case with sections for the RCS High Flow Microbial Air Sampler, two rotors, power supply, RS232 serial cable, USB Adapter, documents and agar strips.

Legal Information

RCS is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service