Skip to Content
Merck
All Photos(1)

Documents

1.17159

Millipore

MAS-100 Iso NT® IP54 Kit

for use with MAS-100 Iso NT®, to protect the input side of air samplers from dust and water sprays

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122409
NACRES:
NA.74

material

aluminum covering

Agency

non-compendial

feature

installed

packaging

pkg of 1 kit

application(s)

air monitoring
pharmaceutical

compatibility

for use with MAS-100 Iso NT®

storage temp.

no temp limit

General description

IP54 Kit is an additional accessory designed to be specifically used for MAS-100 ISO NT® air sampler. The MAS-100 ISO NT® is used for microbial monitoring at fixed sampling locations in isolators. The kit is to protect the input side of air samplers from dust and indirect water sprays.

Application

MAS-100 ISO NT® IP54 Kit is an additional accessory used in protecting the instrument from dust and indirect water sprays. The MAS-100 ISO NT® is extensively used in Pharmaceutical applications.

Legal Information

MAS-100 ISO NT is a registered trademark of MBV AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service