Skip to Content
Merck
All Photos(1)

Documents

1.11615

Sigma-Aldrich

Ethidium bromide

for biochemistry

Synonym(s):

Ethidium bromide, 5-Ethyl-3,8-diamino-6-phenylphenanthridinium bromide, Homidium bromide, EtBr

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
394.31
MDL number:
UNSPSC Code:
41105322
EC Index Number:
214-984-6

Quality Level

form

solid

potency

1503 mg/kg LD50, oral (Rat)

pH

4.4 (20 °C, 10 g/L in H2O)

mp

261-264 °C

transition temp

flash point >100 °C

solubility

40 g/L

density

1.01 g/cm3 at 20 °C

bulk density

340 kg/m3

storage temp.

2-30°C

InChI

1S/C21H19N3.BrH/c1-2-24-20-13-16(23)9-11-18(20)17-10-8-15(22)12-19(17)21(24)14-6-4-3-5-7-14;/h3-13,23H,2,22H2,1H3;1H

InChI key

ZMMJGEGLRURXTF-UHFFFAOYSA-N

General description

for biochemistry

Application

Ethidium bromide for biochemistry. CAS 1239-45-8, pH 4.4 (10 g/l, H₂O, 20 °C).

Analysis Note

Assay (perchloric acid titration, calculated on dried substance)≥ 98 %Identity (UV/VIS-Spectrum)passes testAbsorption maximum λmax. (methanol)524 - 527 nmSpec. Absorptivity A 1%/1cm (λmax; 0.05 g/l; methanol; calc. on dried substance)155 - 170TLC-Testpasses testLoss on drying (130 °C, 4 h)≤ 8 %

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 4 Oral - Muta. 2

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service