Skip to Content
Merck
All Photos(1)

Documents

1.10620

Millipore

LMX broth modified acc. to MANAFI and OSSMER

for microbiology Fluorocult®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.74

Quality Level

sterility

non-sterile

form

granular

manufacturer/tradename

FluoroCult®

technique(s)

microbiological culture: suitable

pH

6.6-7.0 (25 °C, 17 g/L in H2O, after autoclaving)

solubility

17 g/L

application(s)

food and beverages
microbiology

storage temp.

15-25°C

suitability

Escherichia coli
coliforms

Legal Information

FLUOROCULT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Selective media enable faster results and visual confirmation for the detection, identification, and enumeration of microorganisms

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service