Skip to Content
Merck
All Photos(1)

Documents

03-0123-00

Millipore

SMC® Mouse IL-17A Discovery Immunoassay Kit

Synonym(s):

mouse IL 17A immunoassay kit, mouse IL-17A Single Molecule Counting Kit, mouse IL-17A ultra-sensitive immunoassay, mouse Interleukin 17A immunoassay kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.47

species reactivity

mouse

Quality Level

manufacturer/tradename

SMC®

assay range

accuracy: 80-120%
(minimum of 10 samples spiked with acceptable recovery)

technique(s)

ELISA: suitable
single molecule counting: suitable

General description

Interleukin-17A (IL-17A) is the founding member of a group of cytokines called the IL-17 family. There are several members of the IL-17 family, from IL-17A to IL-17F (IL-17E is also called IL-25). IL-17A is involved in mediating proinflammatory responses by inducing the production of many other cytokines, chemokines, and prostaglandins from many cell types. The increased expression of chemokines attracts other cells, including neutrophils. IL-17A has many physiological functions, such as airway remodeling. As a result of these roles, the IL-17 family has been linked to many immune/autoimmune related diseases, including rheumatoid arthritis (RA), asthma, allergic responses, lupus, allograft rejection and antitumor immunity.
The SMC® Mouse IL-17A Discovery Immunoassay Kit contains all reagents required to perform the assays. This assay takes advantage of a standard immunoassay workflow configured in a standard plate format. Assay protocols are similar to existing sandwich ELISA methods with two key differences: 1) Elution buffer disrupts the sandwich, separating the labeled detection antibody for quantification. 2) The SMC® immunoassay system detects analytes using Single Molecule Counting (SMC®) technology.

Application

Research Category
Inflammation & Immunology
Research Sub Category
Cytokines & Cytokine Receptors
The Single Molecule Counting (SMC®) technology gives the SMC® Mouse IL-17A Discovery Immunoassay Kit unparalleled performance in the quantification of IL-17A in mouse serum.
Used to detect/quantify:IL-17A

Components

Assay Plate
10X PBS
Blocking Buffer
Standard Diluent
Mouse IL-17A Detection Antibody
Mouse IL-17A Standard
SMC® Mouse IL-17A Immunoassay Kit Instructions
10X Wash Buffer
Buffer C
Elution Buffer B

Analysis Note

Low-end CV% Range: 0-20%|Low-end CV% Average: 11%

Legal Information

SMC is a registered trademark of Singulex

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Repr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service