Skip to Content
Merck
All Photos(2)

Key Documents

857461P

Avanti

16:1 aldehyde-d5

(2E)-hexadecenal(d5), powder

Synonym(s):

110837

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
RM 1,522.00

RM 1,522.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
RM 1,522.00

About This Item

Empirical Formula (Hill Notation):
C16H25OD5
CAS Number:
Molecular Weight:
243.44
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

RM 1,522.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (857461P-1mg)
pkg of 1 × 5 mg (857461P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 857461P

lipid type

sphingolipids
deuterated lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[2H]C([2H])([2H])C([2H])([2H])CCCCCCCCCCC/C=C/C([H])=O

InChI

1S/C16H30O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17/h14-16H,2-13H2,1H3/b15-14+/i1D3,2D2

InChI key

KLJFYXOVGVXZKT-AKCNZVNDSA-N

General description

Fatty aldehydes are enzymatically converted to fatty alcohols and fatty acids. When assessing S1P lyase enzymatic activity, the estimation of fatty aldehyde stability during the reaction is required. The (2E)-hexadecenal(d5) (857461) is suitable as the internal standard for LC/MS/MS approaches to quantify the product of S1P degradation by S1P lyase, (2E)-hexadecenal; however, a natural (2E)-hexadecenal (857469) is needed to evaluate its stability during enzymatic reaction. The method describing the details of the use of both (2E)-hexadecenal(d5) and (2E)-hexadecenal for LC/MS/MS-based characterization of S1P lyase reaction is in preparation for publication by Berdyshev et al.Personal communication from Evgeny Berdyshev, Ph.D., Assistant Professor, University of Illinois at Chicago.

Application

16:1 aldehyde-d5 has been used as an internal standard in lipid extraction from lung tissue and liquid chromatography/with tandem mass spectrometry analysis (LC/MS/MS).[1] It has also been used as a standard in lipid extraction from plasma, derivatization of 2E-hexadecenal for quantification in quadrupole/time-of-flight (QTOF) mass spectrometer connected with a rapid-resolution-liquid-chromatography (RRLC).[2]

Biochem/physiol Actions

16:1 aldehyde is a predominant fatty aldehyde in phospholipid of bovine adipose tissue,[3] Lactobacillus[4] and few bacteria like Selenomonas ruminantium, when grown on glucose and even-numbered volatile fatty acids.[5] It is one of the main aldehydes in plasmalogen of notched brittle star, Ophiura sarsi.[6]

Packaging

5 mL Amber Glass Screw Cap Vial (857461P-1mg)
5 mL Amber Glass Screw Cap Vial (857461P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel methods for the quantification of (2E)-hexadecenal by liquid chromatography with detection by either ESI QTOF tandem mass spectrometry or fluorescence measurement
Luth A, et al.
Analytica Chimica Acta, 722(8), 70-79 (2012)
Lipid Metabolism in Ruminant Animals (2014)
Irina Gorshkova et al.
Journal of lipid research, 53(8), 1553-1568 (2012-05-23)
The enforcement of sphingosine-1-phosphate (S1P) signaling network protects from radiation-induced pneumonitis. We now demonstrate that, in contrast to early postirradiation period, late postirradiation sphingosine kinase-1 (SphK1) and sphingoid base-1-phosphates are associated with radiation-induced pulmonary fibrosis (RIF). Using the mouse model
Ether Lipids Chemistry and Biology, 335-335 (2012)
Marine Biogenic Lipids, Fats & Oils, 213-213 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service