Skip to Content
Merck
All Photos(2)

Documents

840492C

Avanti

22:6 PG

Avanti Research - A Croda Brand 840492C

Synonym(s):

1,2-didocosahexaenoyl-sn-glycero-3-[phospho-rac-(1-glycerol)] (sodium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C50H74O10PNa
CAS Number:
Molecular Weight:
889.08
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (840492C-25mg)
pkg of 1 × 4 mL (840492C-100mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840492C

concentration

10 mg/mL (840492C-25mg)
25 mg/mL (840492C-100mg)

shipped in

dry ice

storage temp.

−20°C

InChI

1S/C50H75O10P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-37-39-41-49(53)57-45-48(46-59-61(55,56)58-44-47(52)43-51)60-50(54)42-40-38-36-34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h5-8,11-14,17-20,23-26,29-32,35-38,47-48,51-52H,3-4,9-10,15-16,21-22,27-28,33-34,39-46H2,1-2H3,(H,55,56);/q;+1/p-1/b7-5-,8-6-,13-11-,14-12-,19-17-,20-18-,25-23-,26-24-,31-29-,32-30-,37-35-,38-36-;/t47?,48-;/m1./s1

InChI key

LUJLIUULIHKZSH-LJVBEQEHSA-M

General description

22:6 PG or 1,2-didocosahexaenoyl-sn-glycero-3-[phospho-rac-(1-glycerol)] is a sodium salt of glycerophospholipid, with substitution of two chains of docosahexaenic acid at the sn-1 and sn-2 positions and phospho-rac-(1-glycerol) at the sn-3 position of the glycerol backbone.

Application

22:6 PG or 1,2-didocosahexaenoyl-sn-glycero-3-[phospho-rac-(1-glycerol)] (sodium salt) has been used as a lipid standard to optimize the liquid chromatography mass spectroscopy (LC-MS) method conditions.

Packaging

5 mL Clear Glass Sealed Ampule (840492C-100mg)
5 mL Clear Glass Sealed Ampule (840492C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Liver,Kidney, Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mónica Narváez-Rivas et al.
Journal of chromatography. A, 1440, 123-134 (2016-03-02)
The goal of untargeted lipidomics is to have high throughput, yet comprehensive and unambiguous identification and quantification of lipids. Novel stationary phases in LC separation and new mass spectrometric instruments capable of high mass resolving power and faster scanning rate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service