W379107
Octahydrocoumarin
≥98% (mixture of cis and trans)
Synonym(s):
Bicyclononalactone
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C9H14O2
CAS Number:
Molecular Weight:
154.21
FEMA Number:
3791
EC Number:
MDL number:
UNSPSC Code:
12164502
Flavis number:
13.161
Recommended Products
Assay
≥98% (mixture of cis and trans)
refractive index
n20/D 1.492 (lit.)
bp
145 °C/16 mmHg (lit.)
density
1.092 g/mL at 25 °C (lit.)
SMILES string
O=C1CCC2CCCCC2O1
Biochem/physiol Actions
Odor at 1.0%
Taste at 100 ppm
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
177.8 °F - closed cup
Flash Point(C)
81 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Mosciano, G.
Perfumer & Flavorist, 27, 71-71 (2002)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service