Skip to Content
Merck
All Photos(1)

Key Documents

W205907

Sigma-Aldrich

Amyl butyrate

≥99%

Synonym(s):

Pentylbutyrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H18O2
CAS Number:
Molecular Weight:
158.24
FEMA Number:
2059
Council of Europe no.:
270c
MDL number:
Flavis number:
9.044

Assay

≥99%

refractive index

n20/D 1.41 (lit.)

bp

184-188 °C (lit.)

density

0.863 g/mL at 25 °C (lit.)

SMILES string

CCCCCOC(=O)CCC

InChI

1S/C9H18O2/c1-3-5-6-8-11-9(10)7-4-2/h3-8H2,1-2H3

InChI key

CFNJLPHOBMVMNS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Banana, pineapple, cherry, sweet

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service