Skip to Content
Merck
All Photos(1)

Key Documents

GF59489814

Havar® - high-strength non-magnetic alloy, Co42.5Cr20Ni13FeWMoMn

foil, 8mm disks, 0.05mm thickness, as rolled

Synonym(s):

CO010270

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11101712
NACRES:
NA.23

form

foil (disks - as rolled)

manufacturer/tradename

Goodfellow 594-898-14

size × thickness

8 mm × 0.05 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Legal Information

Havar is a registered trademark of Precision Metals, Inc. AMETEK, or its subsidiaries or affiliates.
Havar is a registered trademark of Precision Metals, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service