Skip to Content
Merck
All Photos(1)

Key Documents

DWK217211706

Duran® Round Bottom Flask

narrow-mouth neck, WITH BEADED RIM

Synonym(s):

217211706

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121804
NACRES:
NB.43

material

narrow-mouth neck
round bottom

packaging

pkg of 10 ea

manufacturer/tradename

DURAN® 217211706

capacity

50 mL

Looking for similar products? Visit Product Comparison Guide

Application

Uniform wall thickness distribution makes these flasks ideal for heating applications. Size 2000, 3000, 5000 ml non-DIN EN ISO size., Size 12000 and 20000 not DIN ISO, they are ASTM E 1403 Boro 3.3

Features and Benefits

The geometry permits very uniform heating. Fired-on,

Legal Information

DURAN is a registered trademark of DWK Life Sciences

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service