Skip to Content
Merck
All Photos(1)

Key Documents

D38200

Sigma-Aldrich

Dibromoacetonitrile

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br2CHCN
CAS Number:
Molecular Weight:
198.84
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

95%

refractive index

n20/D 1.539 (lit.)

bp

67-69 °C/24 mmHg (lit.)

density

2.296 g/mL at 25 °C (lit.)

SMILES string

BrC(Br)C#N

InChI

1S/C2HBr2N/c3-2(4)1-5/h2H

InChI key

NDSBDLSWTGLNQA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Irrit. 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A E Ahmed et al.
Journal of biochemical toxicology, 6(2), 115-121 (1991-01-01)
The haloacetonitrile, dibromoacetonitrile (DBAN), is a direct-acting genotoxic agent that has been detected in drinking water. In a time course study, male Sprague-Dawley rats were treated with DBAN (75 mg/kg PO), and killed at 0.5, 1, 2, and 4 hr
Mohamed H Abdel-Wahab
Pharmacological research, 47(6), 509-515 (2003-05-14)
Dibromoacetonitrile (DBAN) is known to be water disinfectant by-product. Its broad-spectrum toxicity in different test systems in vivo and in vitro has been reported. Oxidative damage induced by DBAN may be partially responsible for its toxicity. Herein, the ability of
Ahmed M Mohamadin et al.
Archives of toxicology, 77(2), 86-93 (2003-02-19)
Dibromoacetonitrile (DBAN) is a disinfection byproduct of chlorination of drinking water. Epidemiological studies indicate that it might present a potential hazard to human health. The present work provides evidence for DBAN activation to cyanide (CN(-)) by the hypoxanthine (HX)/xanthine oxidase
R Poon et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 41(8), 1051-1061 (2003-07-05)
The subchronic toxicity of dibromoacetonitrile (DBAN), a disinfection by-product in drinking water, was studied in the rat. Male (180+/-18 g) and female (152+/-9 g) Sprague-Dawley rats (10 animals per group) were fed DBAN in organic-free distilled water at concentrations of
Dibromoacetonitrile.
IARC monographs on the evaluation of carcinogenic risks to humans, 71 Pt 3, 1369-1373 (1999-09-07)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service