Skip to Content
Merck
All Photos(1)

Key Documents

B62005

Sigma-Aldrich

2-Bromo-4-chlorophenol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H3(Cl)OH
CAS Number:
Molecular Weight:
207.45
Beilstein:
2042871
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

bp

121-123 °C/10 mmHg (lit.)

mp

31-33 °C (lit.)

SMILES string

Oc1ccc(Cl)cc1Br

InChI

1S/C6H4BrClO/c7-5-3-4(8)1-2-6(5)9/h1-3,9H

InChI key

ZIYRDJLAJYTELF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tarek Manasfi et al.
International journal of hygiene and environmental health, 222(1), 1-8 (2018-07-23)
Chlorination of seawater is one of the most effective technologies for industrial biofouling control. However, chlorination leads to the formation of halogenated chlorination byproducts (CBPs) associated with potential risks to environmental and human health. The present study investigated the occurrence

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service