78664
(R)-Boc-3-(2-naphthyl)-β-Ala-OH
≥98.0% (HPLC)
Synonym(s):
(R)-3-(Boc-amino)-3-(2-naphthyl)propionic acid, L-Boc-3-(2-naphthyl)-β-alanine
About This Item
Assay
≥98.0% (HPLC)
form
powder
color
white
mp
158 °C
application(s)
peptide synthesis
SMILES string
CC(C)(C)OC(=O)N[C@H](CC(O)=O)c1ccc2ccccc2c1
InChI
1S/C18H21NO4/c1-18(2,3)23-17(22)19-15(11-16(20)21)14-9-8-12-6-4-5-7-13(12)10-14/h4-10,15H,11H2,1-3H3,(H,19,22)(H,20,21)/t15-/m1/s1
InChI key
NCQJBPXXRXOIJD-OAHLLOKOSA-N
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service