Skip to Content
Merck
All Photos(1)

Key Documents

775207

Sigma-Aldrich

N-(4-Bromophenyl)maleimide

97%

Synonym(s):

1-(4-Bromophenyl)-1H-pyrrole-2,5-dione, N-(p-Bromophenyl)maleimide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H6BrNO2
CAS Number:
Molecular Weight:
252.06
UNSPSC Code:
12352100
NACRES:
NA.22

Assay

97%

form

powder

mp

125-130 °C

InChI

1S/C10H6BrNO2/c11-7-1-3-8(4-2-7)12-9(13)5-6-10(12)14/h1-6H

InChI key

FECSFBYOMHWJQG-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

E J Breaux et al.
Biomedical & environmental mass spectrometry, 15(3), 123-128 (1988-02-01)
A new mass spectral method has been developed and used to identify biologically important thiols. This method is based upon the use of C-14 labeled N-para-bromophenylmaleimide (BPM) to selectively derivatize thiols. The isotopic label facilitates the isolation and purification of
Andrea Forgacsova et al.
Journal of pharmaceutical and biomedical analysis, 164, 442-451 (2018-11-18)
An ultra-high performance hydrophilic interaction liquid chromatography - triple quadrupole tandem mass spectrometry method was developed for the determination of biologically important thiols, namely cysteine, homocysteine, cysteinyl-glycine, glutathione, in rat plasma. The sample preparation procedure as well as the analytical

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service