Skip to Content
Merck
All Photos(1)

Documents

638854

Sigma-Aldrich

7-Nitroindole

97%

Synonym(s):

NSC 69874

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6N2O2
CAS Number:
Molecular Weight:
162.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

94-98 °C (lit.)

SMILES string

[O-][N+](=O)c1cccc2cc[nH]c12

InChI

1S/C8H6N2O2/c11-10(12)7-3-1-2-6-4-5-9-8(6)7/h1-5,9H

InChI key

LZJGQIVWUKFTRD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for preparation of:
  • Protein kinase inhibitors
  • Potential fructose 1,6-bisphosphatase inhibitors
  • Factor Xa inhibitors
  • Antagonist of the mineralocorticoid receptor
  • Antitumor sulfonamides
  • Inosine monophosphate dehydrogenase (IMPDH) inhibitors
  • Subtype-selective cyclooxygenase (COX) inhibitors
  • Thrombin protease-activated receptor (PAR-1) ligands

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chaitany Jayprakash Raorane et al.
Biomolecules, 10(8) (2020-08-23)
Multi-drug resistant Acinetobacter baumannii is well-known for its rapid acclimatization in hospital environments. The ability of the bacterium to endure desiccation and starvation on dry surfaces for up to a month results in outbreaks of health care-associated infections. Previously, indole

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service