Skip to Content
Merck
All Photos(1)

Key Documents

592064

Sigma-Aldrich

Poly(pentabromophenyl methacrylate)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C10H5Br5O2)n
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

refractive index

n20/D 1.710 (lit.)

transition temp

Tg (DSC) 37 °C (onset)

solubility

chloroform: soluble

Application

High refractive index polymer for optical waveguides

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Polymeric materials, with unique refractive index properties, find applications in photonics and optical coatings for various devices.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service