Skip to Content
Merck
All Photos(1)

Key Documents

589845

Sigma-Aldrich

Boc-Met-OH-1-13C

99 atom % 13C

Synonym(s):

N-(tert-Butoxycarbonyl)-L-methionine-1-13C, L-Methionine-1-13C, N-t-Boc derivative

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SCH2CH2CH(NH-Boc)13CO2H
CAS Number:
Molecular Weight:
250.32
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

isotopic purity

99 atom % 13C

form

solid

technique(s)

bio NMR: suitable

mp

47-50 °C (lit.)

application(s)

peptide synthesis

functional group

Boc

mass shift

M+1

SMILES string

CSCC[C@H](NC(=O)OC(C)(C)C)[13C](O)=O

InChI

1S/C10H19NO4S/c1-10(2,3)15-9(14)11-7(8(12)13)5-6-16-4/h7H,5-6H2,1-4H3,(H,11,14)(H,12,13)/t7-/m0/s1/i8+1

InChI key

IMUSLIHRIYOHEV-BAZIHRRBSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service